Mangan 1.000 mg/l i utspädd salpetersyra (av Mn(OAc)2) ARISTAR® standard för ICP
Leverantör: VWR Chemicals
455642FEA 1700 SEK
455642F 455644H
Mangan 1.000 mg/l i utspädd salpetersyra (av Mn(OAc)2) ARISTAR® standard för ICP
Standarder Cationic Standards Manganese Standards
Mn(C₂H₃O₂)₂ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.