Bly 1.000 mg/l i utspädd salpetersyra (av Pb(NO3)2) ARISTAR® standard för ICP
Leverantör: VWR Chemicals
455562GEA 1650 SEK
455562G 455564Y
Bly 1.000 mg/l i utspädd salpetersyra (av Pb(NO3)2) ARISTAR® standard för ICP
Standarder Cationic Standards Lead Standards
Pb(NO₃)₂ in 2% HNO₃.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.