Aluminium standardlösningar för ICP, ARISTAR®
Leverantör: VWR Chemicals
455002CEA 1600 SEK
455002C 455012E 455014G 455004E
Aluminium standardlösningar för ICP, ARISTAR®
Standarder Cationic Standards Aluminium Standards
Al(NO₃)₃.9H₂O in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
Supplied with certificate of analysis.
Certifikat: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Leveransinformation: Supplied with Certificate of Analysis.