Krom 99,99 Metaller basis, powder -60 mesh

Leverantör: Thermo Fisher Scientific

44556.22 44556.36
44556.22EA 3275 SEK
44556.22 44556.36
Krom 99,99 Metaller basis, powder -60 mesh
Krom

Purity: 99.99% CAS No.: 7440-47-3 Odour: Odourless MDL No.: MFCD00010944

Formel: Cr
MW: 52 g/mol
Kokpunkt: 2469
Smältpunkt: 1840
Densitet: 7.2
Storage Temperature: Omgivande
MDL Number: MFCD00010944
CAS nummer: 7440-47-3
EINECS: 231-157-5
UN: 3089
ADR: 4.1 III
Merck Index: 12,02288

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR