STYRENE atactic, M.W. 125,000-250,000

Leverantör: Thermo Fisher Scientific

44537.09 44537.18
44537.09EA 895 SEK
44537.09 44537.18
STYRENE atactic, M.W. 125,000-250,000
Poly(styrene)

CAS No.: 9003-53-6 Formula Weight: 125,000-250,000 Physical Form: Beads Odour: Odourless MDL No.: MFCD00084450

Formel: C₈H₈
MW: 125000 - 250000 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00084450
CAS nummer: 9003-53-6

Order Now

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR