Natriumdodecylsulfat 100 g/l aqueous solution, Electran Electrophoresis
Leverantör: VWR Chemicals
Fara
Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
444062FEA 865 SEK
444062F
Natriumdodecylsulfat 100 g/l aqueous solution, Electran Electrophoresis
Natriumdodecylsulfat
Prepared from 10807 SDS AnalaR NORMAPUR®
Formel: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Densitet: 1…1.1 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00036175
CAS nummer: 151-21-3
EINECS: 205-788-1
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Assay (W/V) (at preparation) 9.8 - 10.2 %