VWR®, Rocking platform shaker
Leverantör: VWR Collection
444-0756EA 11550 SEK
444-0756 444-0757 444-0758
VWR®, Rocking platform shaker
Skakapparater Skakapparater, roterande/vaggande rörelse
Rocking platform shaker with low profile design can be used in most fume hoods and incubators. The cast aluminium base offers durability and stability, the spill resistant design channels fluids away from internal components. Precise speed control provides smooth, low speed rocking motion down to one min⁻¹.
  • Microprocessor control provides electronic tilt angle adjustment from zero to 15° while unit is operating
  • LED display shows speed, tilt angle and time simultaneously
  • Timer from 1 s to 160 h, with audible alarm when the time reaches zero
  • Can be used at –1
More Product Information