Natriumhydroxid 0.1 mol/l (0.1 N) aqueous solution, Supelco®

Leverantör: Merck

Synonyms: Natronlut, Kaustik soda

MQ100

43617-1L
43617-1LEA 591 SEK
43617-1L
Natriumhydroxid 0.1 mol/l (0.1 N) aqueous solution, Supelco®
Natriumhydroxid
Formel: NaOH
MW: 40 g/mol
Kokpunkt: 1390
Smältpunkt: 323
Densitet: 1 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003548
CAS nummer: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 III
Merck Index: 14,08627
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e121c7c6d29f20a905f9749f2ef67288f60e2a92&vid=f0a6855bd87c524a863a423fd6906a12923501d7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR