VWR®, Bead Mill Homogeniser
Leverantör: VWR Collection
432-0367EA 0 SEK
432-0367
VWR®, Bead Mill Homogeniser
Homogenisatorer Homogenisatorer, bordsmodeller
The VWR® Bead Mill homogeniser is a part of the newest generation of Bead Mill homogenisers. This economical system is specifically designed for grinding, lysing and homogenisation of up to 16 samples. Using sample tubes pre-filled with a variety of lysing beads, the VWR Bead Mill vigorously and uniformly shakes the tubes providing an efficient, consistent high yield and quality homogenisation usually in less than 40 seconds.
  • Capacity for 16 samples (12×2 ml tubes plus 4×7 ml tubes, 16 in total)
  • Run time: 1 second to 10 minutes
  • 0,8 to 6 m/s
  • Up to 99
More Product Information