Ultrasonic processors, Vibra-Cell™ VCX 130 and VCX 130PB
Leverantör: SONICS MATERIALS
VCX130 VCX-130-220
432-0135EA 0 SEK
432-0135 SONIVCX-130-220
Ultrasonic processors, Vibra-Cell™ VCX 130 and VCX 130PB
Homogenisatorer Desintegratorer, ultraljud
The Vibra-Cell™ is a technologically advanced, high-intensity ultrasonic processor for low volume applications*. It can safely process a wide variety of organic and inorganic materials in applications such as cell disruption, sample preparation, homogenisation, disaggregation and sonochemical reactions. The system can vary the power output and constantly monitor both power (Watts) and energy (Joules). The VCX 130 model has the additional features of 10 hour timer, independent on-off pulser and ability to program some set and run parameters.
  • Automatic tuning eliminates need for constant
More Product Information