Disperser system, ULTRA-TURRAX® tube drive P control
Leverantör: IKA
25005981
431-0317EA 21350 SEK
431-0317
Disperser system, ULTRA-TURRAX® tube drive P control
Homogenisatorer Homogenisatorer, bordsmodeller
Universal disposable disperser system with disposable sample tubes that can be hermetically sealed. Provides protection and security from infectious sample materials, toxic substances or substances with strong odours.
  • Sealable disposable tubes optimised for your application
  • Tubes reduce the amount of cleaning required and rule out any risk of cross-contamination
  • Programmable test conditions
  • Adjustable change in direction of rotation
  • Turbo button
Programmable test conditions - user can specify and systematically program the test conditions and storeMore Product Information
Order Now
SPECIFIKATIONER
Modell Tube Drive P control disperser
Environmentally Preferable
Volym, H₂O 2 - 15 ml (ST-20 mixing/BMT-20 crushing) Modes: A, B, P1-P9
5 - 15 ml (DT-50) Modes: A, B, P1-P9
15 - 50 ml (ST-50 stirring/DT-50 stirring/BMT-50 wet/dry mixing/DIS 50 wet mixing/dissolving) Modes: Turbo, Rev, A, B, P1-P9
100 - 300 ml (DIS 300) Modes: Turbo
Max. viskositet 5000 mPas
Hastighetsområde 400 - 8000 min⁻¹ (Turbo speed)
≤600 min⁻¹ (Mode A and B)
≤600 min⁻¹ (Mode P1-P9)
Hastighetsstabilitet 3%
Effekt in/ut 36/28 W
Tidsvisning Digital with OLED display
Timer 10 s - 30 min (infinitely adjustable)
Display Digital with OLED display
B×D×H 122×178×54 mm
Vikt 1,3 kg
Sound level 50 dB(A)