Natrium standard dissolved in oil, 5.000 µg/g Na in mineral oil Specpure® for wear metal analysis

Leverantör: Thermo Fisher Scientific

42449.18
42449.18EA 2450 SEK
42449.18
Natrium standard dissolved in oil, 5.000 µg/g Na in mineral oil Specpure® for wear metal analysis
Standarder Cationic Standards Sodium Standards

Physical Form: Liquid
MDL No.: MFCD00085307

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR