Urea 99,5, ultrapure_bioreagent, J.T.Baker®

Leverantör: Avantor
Varning

Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide

4111-01EA 0 SEK
4111-01 4111-05 4111-07 4111-F9
Urea 99,5, ultrapure_bioreagent, J.T.Baker®
Urea

For protein solubilization and denaturation. DNase-, RNase-, protease-, and cyanate-free.

Formel: NH₂CONH₂
MW: 60.06 g/mol
Kokpunkt: 197
Smältpunkt: 132.7
Densitet: 1.34 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00008022
CAS nummer: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 13,09935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

For Laboratory,Research,or Manufacturing Use ≥99.5%
Assay (NH₂CONH₂) 98.0 - 102.0 %
Appearance Passes Test
RNase Activity Passes Test
DNase Activity (None Detected) Passes Test
Protease Activity Passes Test
Absorbance of a 5M Solution - 260 nm ≤0.050
Absorbance of a 5M Solution - 280 nm ≤0.030
Biuret ≤0.01
Cyanate (CNO) None Detected
Insoluble Matter ≤0.005 %
Water (H₂O)(by Karl Fischer titrn) ≤0.5 %
Conductivity of 8.5M Solution at 25°C, (µmho/cm) ≤30
Chloride (Cl) ≤1 ppm
Trace Impurities - Copper (Cu) ≤1 ppm
Trace Impurities - Iron (Fe) ≤1 ppm
Trace Impurities - Lead (Pb) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR