Copper Standard for IC, TraceCERT®, Supelco®
Leverantör: Merck
40786-100ML
40786-100MLEA 638 SEK
40786-100ML
Copper Standard for IC, TraceCERT®, Supelco®
Standarder Cationic Standards Copper Standards
Certifikat: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034