RedDot™1 Far-Red Nuclear Stain, 200X in Water, Biotium
Leverantör: Biotium
40060-1 40060-T 40060
40060-1EA 8025 SEK
40060-1 40060-T 40060.
RedDot™1 Far-Red Nuclear Stain, 200X in Water, Biotium
Analyser Cellulär analys
RedDot™1 is a far-red cell membrane-permeable nuclear dye similar to Draq5™. The dye is ideal for specifically staining the nuclei of live cells.
  • Ideal for specifically staining the nuclei of live cells for short term imaging
  • Highly thermostable and photostable, for convenient handling and demanding imaging applications
  • Can be used for DNA content analysis by flow cytometry like Vybrant™ DyeCycle™ Ruby
  • Can be used for normalization of In Cell Western® in fixed cells, similar to Draq5™/Sapphire®700
  • λEx/λEm = 662/694 nm (with DNA), for detection in the Cy®5 channel
RedDot™1 can be excited at a wide range of wavelengths between 488 nm and 647 nm, including the 488 nm laser for flow cytometry, and can be used for DNA content analysis by flow cytometry. While DAPI and Hoechst dyes show a strong preference for A-T rich regions, RedDot™1 is relatively insensitive to sequence base composition.
Like Draq5®, RedDot™1 is toxic to cells within a few hours, and is recommended for short term staining experiments. For a low-toxicity far-red nuclear stain for live cells, see our NucSpot™ 650 dye.
RedDot™1 cannot be used for nuclear-specific staining in fixed cells. Please see RedDot™2 (cat# 40061), a spectrally similar dye designed for specific nuclear staining of fixed and permeabilized cells or selective nuclear staining of dead cells.
Please note: far-red dyes like RedDot™1 are not visible to the human eye, but must be imaged with a CCD camera or by confocal microscopy.
Cy dye is a registered trademark of GE Healthcare. Draq5 is a trademark of BioStatus Ltd. In Cell Western and Sapphire are registered trademarks of LI-COR Inc. Vybrant and DyeCycle are trademarks of Thermo Fisher Scientific.