Titan standardlösning, 10000 mg/l Ti in 5% salpetersyra with hydrofluoric acid (max. 1%) (av Ti) Specpure® standard för ICP

Leverantör: Thermo Fisher Scientific

35759.ADEA 2100 SEK
35759.AD 35759.AE 35759.AP 35768.AE 38720.AD 38720.AE 38725.AP 35768.AD 35768.AP 38720.AP 38725.AD 38725.AE 45267.AE
Titan standardlösning, 10000 mg/l Ti in 5% salpetersyra with hydrofluoric acid (max. 1%) (av Ti) Specpure® standard för ICP
Standarder Cationic Standards Titanium Standards

UN No.: 3264
MDL No.: MFCD00011264


Certifikat: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR