Kaliumpermanganat concentrate for dilution to 1L, 1/500 M KMnO4 (0.01N) Volumetric, Fluka™

Leverantör: Honeywell Chemicals

Synonyms: Permanganic acid potassium salt

38136-1EA
38136-1EAEA 955 SEK
38136-1EA
Kaliumpermanganat concentrate for dilution to 1L, 1/500 M KMnO4 (0.01N) Volumetric, Fluka™
Kaliumpermanganat

Concentrate for 1L standard solution

Formel: KMnO₄
MW: 158.03 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00011364
CAS nummer: 7722-64-7
EINECS: 231-760-3
Merck Index: 12,07824

Order Now

Specifikation testresultat

Factor 0.998 - 1.002
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR