Bensoesyra 99,5 ACS

Leverantör: Thermo Fisher Scientific
Fara

36230.14 36230.22 36230.36
36230.14EA 655 SEK
36230.14 36230.22 36230.36
Bensoesyra 99,5 ACS
Bensoesyra

Purity: 99.5% min CAS No.: 65-85-0 Molecular Formula: C7H6O2 Formula Weight: 122.12 Flash Point: 121°(250°F) Physical Form: Crystalline Density: 1.32 Appearance: White Odour: Weak, characteristic pH: 2.5-3.5 Melting Point: 121-124° Boiling Point: 249° Beilstein Reference: 636131 Merck Reference: 14,1091 MDL No.: MFCD00002398

Formel: C₇H₆O₂
MW: 122.12 g/mol
Kokpunkt: 249 °C
Smältpunkt: 121…124 °C
Densitet: 1.32
Flampunkt: 121 °C (250 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00002398
CAS nummer: 65-85-0
EINECS: 200-618-2
REACH: 01-2119455536-33
Merck Index: 14,1091
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d561f311b11ea8ab2c8f07253e6748e588eb71aa&vid=dcf5e45072cda5106216b497cd29716270ae772c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR