Supelco® PESTANAL® Organic Reference Standard, Mesosulfuron-Methyl
Leverantör: Merck
34178-100MG
34178-100MGEA 1210 SEK
34178-100MG
Supelco® PESTANAL® Organic Reference Standard, Mesosulfuron-Methyl
Standarder Organic Standards
  • GHS signal: GHS09
Observera: P273 - P391 - P501
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9bb54c23a95835486a07f85a69cff4824cc996cd&vid=61c226c7851e59f7486912acc72fdef6fb586589&prid=web&clid=SAPDAM
Order Now
SPECIFIKATIONER
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 208465-21-8
MDL No. MFCD04112616
Formel C₁₇H₂₁N₅O₉S₂
Molekylvikt 503,51
Faroklassbeskrivning H410
Flampunkt Not applicable
Shipping conditions STD
Storage conditions 2...30 °C