Supelco® PESTANAL® Organic Reference Standard, Metolachlor OA

Leverantör: Merck

34148-10MG
34148-10MGEA 3010 SEK
34148-10MG
Supelco® PESTANAL® Organic Reference Standard, Metolachlor OA
Standarder Organic Standards
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51a1b9facfe5b59ca4913711d9d51f9d6500e967&vid=51a1b9facfe5b59ca4913711d9d51f9d6500e967&prid=web&clid=SAPDAM

Order Now

SPECIFIKATIONER

Synonym(s) BSTFA; TMCS; N-(2-Ethyl-6-methylphenyl)-N-(2-methoxy-1-methylethyl)oxalamic acid; [(2-Ethyl-6-methylphenyl)(2-methoxy-1-methylethyl)amino]oxo-acetic acid
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 152019-73-3
MDL No. MFCD08702643
Formel C₁₅H₂₁NO₄
Molekylvikt 279,33
Flampunkt Not applicable
Shipping conditions STD
Storage conditions 2...30 °C
Förvaringstemperatur

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR