Organic reference standard, Deoxynivalenol solution, analytical standard, Supelco®
Leverantör: Merck
34124-2ML
34124-2MLEA 4200 SEK
34124-2ML
Organic reference standard, Deoxynivalenol solution, analytical standard, Supelco®
Standarder Organic Standards
Deoxynivalenol belongs to the group of tetracyclic sesquiterpenoid mycotoxins called trichothecenes, produced by Fusarium fungi, naturally occurring in small grains and infected corn.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2b7afdd856fe1d840baf9e608e00fdcfa42abd6&vid=e2b7afdd856fe1d840baf9e608e00fdcfa42abd6&prid=web&clid=SAPDAM
Order Now
SPECIFIKATIONER
Benämning Deoxynivalenol solution
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 51481-10-8
Formel C₁₅H₂₀O₆
Förvaringstemperatur –20 °C