Aflatoxin Mix, analytical standards for chromatography, Supelco®
Leverantör: Merck
34036-1ML-R 34036-2ML-R
34036-1ML-REA 3560 SEK
34036-1ML-R 34036-2ML-R
Aflatoxin Mix, analytical standards for chromatography, Supelco®
Standarder Organic Standards
These analytical standards are suitable for HPLC and gas chromatography. Mutil component solution with a concentration of 0,5 μg/ml B2 and G2 in acetonitrile and 2 μg/ml B1 and G1 in acetonitrile.
Order Now
SPECIFIKATIONER
Environmentally Preferable
M-Clarity quality level MQ100