Supelco® PESTANAL® Organic Reference Standard, Bentazon-D₇

Leverantör: Merck

32965-10MG
32965-10MGEA 5900 SEK
32965-10MG
Supelco® PESTANAL® Organic Reference Standard, Bentazon-D₇
Standarder Organic Standards

  • GHS signal: GHS07


Observera: P261 - P273 - P280 - P301 + P312 - P302 + P352 - P305 + P351 + P338

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d606aabc14095d7e86e92653f86d09aa153a956d&vid=b2a5b471d01b61bead174dd68cc4a3aeae5eb575&prid=web&clid=SAPDAM

Order Now

SPECIFIKATIONER

Synonym(s) 3-Isopropyl-d₇-1H-2,1,3-benzothiadiazin-4(3H)-one 2,2-dioxide
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 131842-77-8
MDL No. MFCD09953755
Formel C₁₀D₇H₅N₂O₃S
Molekylvikt 247,32
Faroklassbeskrivning H302 - H317 - H319 - H412
Flampunkt Not applicable
Shipping conditions STD
Storage conditions 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR