Metanol-D4 Isotope_(99.8% D) + 0.03% (v/v) TMS, AcroSeal® NMR

Leverantör: Thermo Fisher Scientific
Fara

Synonyms: Methanol-[²H₄], Hydroxy methane-D4, Methanol-d4, Methyl-D3 alcohol-D, Carbinol-D4, Wood alcohol-D4, Methyl alcohol-D4, Tetradeuteromethanol

325351000
325351000.EA 10900 SEK
325351000.
Metanol-D4 Isotope_(99.8% D) + 0.03% (v/v) TMS, AcroSeal® NMR
Metanol-D4
Formel: D₃COD
MW: 36.01 g/mol
Kokpunkt: 65 °C (760 mmHg)
Smältpunkt: –99 °C
Densitet: 0.888 g/cm³ (20 °C)
Flampunkt: 12 °C
Storage Temperature: Omgivande
MDL Number: MFCD00044637
CAS nummer: 811-98-3
EINECS: 212-378-6
UN: 1230
ADR: 3 II

Order Now

Specifikation testresultat

Appearance Clear colorless Liquid
Water ≤0.05 % (Coulometric)
D-Enrichment ≥99.75 %
TMS Contains 0.03 v/v% TMS

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR