Salpetersyra 1 mol/l (1 N), AVS TITRINORM Volumetric
Leverantör: VWR Chemicals
Fara
Synonyms: Nitric acid solution
30065.291EA 177 SEK
30065.291
Salpetersyra 1 mol/l (1 N), AVS TITRINORM Volumetric
Salpetersyra
Formel: HNO₃
MW: 63.01 g/mol
Kokpunkt: 87
Smältpunkt: -42
Densitet: 1.035 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011349
CAS nummer: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 II
Merck Index: 13,06610
Order Now
Specifikation testresultat
Titer (20°C; real value 0.2 % accuracy) 0.998 - 1.002 mol/l
NIST traceable Confirmed
SPECIFIKATIONER
Environmentally Preferable