Saltsyra 2 mol/l (2 N), AVS TITRINORM Volumetric

Leverantör: VWR Chemicals
Varning

Synonyms: Hydrochloric acid solution, Muriatic acid

30025.293EA 222 SEK
30025.293 30025.362 30025.373 30025.407 30025.320
Saltsyra 2 mol/l (2 N), AVS TITRINORM Volumetric
Saltsyra
Formel: HCl
MW: 36.46 g/mol
Densitet: 1.031 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011324
CAS nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 III

Order Now

Specifikation testresultat

Titer (20°C; real value 0.2 % accuracy) 1.996 - 2.004 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR