Saltsyra 1 mol/l (1 N), AVS TITRINORM Reag. Ph. Eur., Reag. USP, NF volumetrisk lösning

Leverantör: VWR Chemicals
Varning

Synonyms: Hydrochloric acid solution, Muriatic acid

30024.324EA 600 SEK
30024.324 30024.370 30024.404 30024.415 30024.448 30024.290
Saltsyra 1 mol/l (1 N), AVS TITRINORM Reag. Ph. Eur., Reag. USP, NF volumetrisk lösning
Saltsyra
Formel: HCl
MW: 36.46 g/mol
Densitet: 1.02 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00011324
CAS nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 III

Order Now

Specifikation testresultat

Conforms to Reag. Ph.Eur. Passes test
Titer (20°C; real value 0.2 % accuracy) 0.998 - 1.002 mol/l
Conforms to Reag. USP-NF Passes test
NIST traceable Confirmed

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR