Aluminium sample spear with spiral pockets
Leverantör: Sampling Systems
1942A-1000 1942A-1500 1942A-2000 1942A-2500
300-0456EA 4500 SEK
300-0456 300-0457 300-0458 300-0459
Aluminium sample spear with spiral pockets
Provtagare Provtagare, fasta ämnen
The spiral sampling spear is ideal for taking samples of powders, grains and granules at multiple locations.
  • Lightweight aluminium
  • Spiral pocket positions for representative sampling
  • Takes multiple samples along its length
  • Robust design
  • Easy to operate
This sampler is ideal for taking representative composite samples from containers, bulk grain, big bags or silos.
The pockets are positioned in a slight spiral so the bottom pocket open first giving a more representative sample. The lightweight sampler is quick and easy to use. The robust double tubeMore Product Information