Aktivt kol, rods, Norit® RBAA-3, Supelco®

Leverantör: Merck

Synonyms: Carbon activated, Activated carbon, Activated charcoal

29238-1KG
29238-1KGEA 1540 SEK
29238-1KG
Aktivt kol, rods, Norit® RBAA-3, Supelco®
Aktivt kol
Formel: C
MW: 12.01 g/mol
Smältpunkt: 3550
MDL Number: MFCD00133992
CAS nummer: 7440-44-0
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR