Natriumsulfat, vattenfri 98.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytiskt reagens

Leverantör: VWR Chemicals

Synonyms: Disodium sulphate, Bisodium sulphate, Sulphuric acid disodium salt

28114.296EA 591 SEK
28114.296 28114.365 28114.230 28114.460 28114.260
Natriumsulfat, vattenfri 98.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytiskt reagens
Sodium sulphate
Formel: Na₂SO₄
MW: 142.04 g/mol
Kokpunkt: 1429
Smältpunkt: 888
Densitet: 2.7 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003504
CAS nummer: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Assay (dried substance) 98.5 - 101.0 %
Acidity or alkalinity Passes test
Appearance of solution Passes test
Identification (SO4) Passes test
Identification (Na) Passes test
pH (25°C; 5 %) 5.2 - 8.0
Heavy metals (as Pb) Max. 5 ppm
Insolubility in water Max. 100 ppm
Loss on drying (130°C) Max. 0.5 %
Loss on ignition (800°C) Max. 0.5 %
Cl (Chloride) Max. 100 ppm
PO4 (Phosphate) Max. 10 ppm
Total N (Nitrogen) Max. 5 ppm
As (Arsenic) Max. 1 ppm
Ca (Calcium) Max. 50 ppm
Fe (Iron) Max. 5 ppm
K (Potassium) Max. 50 ppm
Mg (Magnesium) Max. 25 ppm
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR