Natriumsulfat, anhydrous 98,5-101,0, powder_fine USP, Ph. Eur.

Leverantör: VWR Chemicals

Synonyms: Disodium sulphate, Bisodium sulphate, Sulphuric acid disodium salt

28105.295EA 805 SEK
28105.295 28105.364 28105.466
Natriumsulfat, anhydrous 98,5-101,0, powder_fine USP, Ph. Eur.
Sodium sulphate

Appearance: White or almost white powder, hygroscopic.


Observera: Not for use as Active Pharmaceutical Ingredient (API)

Formel: Na₂SO₄
MW: 142.04 g/mol
Kokpunkt: 1429
Smältpunkt: 888
Densitet: 2.7 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003504
CAS nummer: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Conforms (see CoA/CoS)
Heavy metals (as Pb) Max. 10 ppm
Residual solvents Unlikely by manuf.process
Residues of metal catalysts or reagents Passes test
As (Arsenic) Max. 2 ppm
Cd (Cadmium) Max. 1 ppm
Co (Cobalt) Max. 1 ppm
Hg (Mercury) Max. 2 ppm
Li (Lithium) Max. 0.6 ppm
Ni (Nickel) Max. 1 ppm
Pb (Lead) Max. 3 ppm
Se (Selenium) Max. 0.5 ppm
V (Vanadium) Max. 1 ppm
Conforms to Ph.Eur./USP Passes test
Ph.Eur. specifications: Max. 1 ppm
Assay (dried substance) [Ph.Eur.] 98.5 --> 101.0 %
Identification A Passes test USP
Identification B Passes test USP
Identification C Passes test Ph.Eur.
Appearance of solution Passes test Ph.Eur.
Acidity or alkalinity Passes test USP
Cl (Chloride) [Ph.Eur.] Max. 450 ppm
Loss on drying (130°C) [Ph.Eur.] Max. 0.5 %
USP specifications:
Assay (calculated on dried basis) [USP] Min. 99.0 %
Loss on drying (105°C; 4 h) [USP] Max. 0.5 %
Cl (Chloride) [USP] Max. 0.02 %
Not for use as Active Pharmaceutical Ingredient (API)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR