Protein G Mag Sepharose
Leverantör: Cytiva
28-9513-79 28-9440-08
28-9513-79EA 1575 SEK
28-9513-79 28-9440-08
Protein G Mag Sepharose
Kulor Magnetiska pärlor
Protein G Mag Sepharose simplifies enrichment of target proteins by immunoprecipitation techniques, protein antigen precipitation from solutions using antibodies, or pull-down applications as an in vitro technique to detect physical interactions.
  • Visible and dense Sepharose based magnetic beads with protein G ligand making it easy to spot and collect bound target protein
  • Non-adherent beads eliminate smearing effects and aggregate formation
  • Can be used without detergents
  • Simple capture of target protein in small or large sample volumes (low microliter to
More Product Information