Ab buffer kit
Leverantör: Cytiva
28-9030-59
28-9030-59EA 1750 SEK
28-9030-59
Ab buffer kit
Buffertar Reningsbuffertar för antikroppar
Ab buffer kit contains prepared buffer concentrates for binding, washing, and elution of IgG according to recommended protocols.
The kit also includes neutralising buffer. Ab buffer kit eliminates time consuming buffer preparation, which allows fast, reproducible, and convenient purification work.
Leveransinformation: Includes: 1×50 ml binding buffer (10X stock solution), 1×15 ml elution buffer (10X stock solution), and 1×25 ml neutralising buffer.