Natriummolybdat(VI)dihydrat ≥99.5%, AnalaR NORMAPUR® analytiskt reagens

Leverantör: VWR Chemicals

Synonyms: Molybdic acid, sodium salt dihydrate, Molybdic acid sodium salt dihydrate, Molybdic acid disodium salt dihydrate, di-Sodium molybdate dihydrate, Sodium molybdate dihydrate, Sodium molybdenum oxide dihydrate

27937.236EA 1500 SEK
27937.236 27937.290
Natriummolybdat(VI)dihydrat ≥99.5%, AnalaR NORMAPUR® analytiskt reagens
Natriummolybdat(VI)dihydrat
Formel: Na₂MoO₄·2H₂O
MW: 241.95 g/mol
Smältpunkt: 687
Densitet: 3.28 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149170
CAS nummer: 10102-40-6
EINECS: 231-551-7
Merck Index: 13,08718

Order Now

Specifikation testresultat

Assay Min. 99.5 %
pH (20°C; 5 %) 7 - 10
Cl (Chloride) Max. 50 ppm
NH4 (Ammonium) Max. 10 ppm
NO3 (Nitrate) Max. 100 ppm
PO4 + AsO4 + SiO4 (as PO4) Max. 10 ppm
SO4 (Sulphate) Max. 50 ppm
Fe (Iron) Max. 10 ppm
Pb (Lead) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR