Sand, TECHNICAL, industrial, washed

Leverantör: VWR Chemicals

Synonyms: Silicium dioxide, Silicium (IV) oxide, Silicon dioxide

27461.298EA 685 SEK
27461.298 27461.360 27461.460
Sand, TECHNICAL, industrial, washed
Sand
Formel: SiO₂
MW: 60.08 g/mol
Smältpunkt: 1710 °C
Densitet: 2.66 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD02100519
CAS nummer: 14808-60-7
EINECS: 238-878-4

Order Now

Specifikation testresultat

Solubility in acid Max. 1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR