Kaliumkromat ≥99.5%, AnalaR NORMAPUR® analytiskt reagens

Leverantör: VWR Chemicals
Fara

26774.236EA 1550 SEK
26774.236 26774.293
Kaliumkromat ≥99.5%, AnalaR NORMAPUR® analytiskt reagens
Kaliumkromat
Formel: K₂CrO₄
MW: 194.19 g/mol
Kokpunkt: 1000
Smältpunkt: 985
Densitet: 2.73 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011368
CAS nummer: 7789-00-6
EINECS: 232-140-5
UN: 3087
ADR: 5.1 (6.1) III
Merck Index: 12,07784

Order Now

Specifikation testresultat

Assay Min. 99.5 %
pH (20°C; 5 %) 8.6 - 9.8
Insolubility in water Max. 50 ppm
Cl (Chloride) Max. 10 ppm
SO4 (Sulphate) Max. 100 ppm
Ca (Calcium) Max. 50 ppm
Cu (Copper) Max. 10 ppm
Fe (Iron) Max. 15 ppm
Na (Sodium) Max. 0.03 %
Pb (Lead) Max. 50 ppm

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR