Diisopropyleter ≥99.0% stabilised, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytiskt reagens

Leverantör: VWR Chemicals
Fara

Synonyms: Isopropyl ether

Stabilization: 10 ppm Stabilised med BHT/Ionol

24900.296EA 655 SEK
24900.296 24900.320
Diisopropyleter ≥99.0% stabilised, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytiskt reagens
Diisopropyleter
Formel: (CH₃)₂CHOCH(CH₃)₂
MW: 102.18 g/mol
Kokpunkt: 68.5 °C (1013 hPa)
Smältpunkt: –86 °C
Densitet: 0.726 g/cm³ (20 °C)
Flampunkt: –28 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008880
CAS nummer: 108-20-3
EINECS: 203-560-6
UN: 1159
ADR: 3 II
Merck Index: 13,05232
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=932b8c021b2bfa9a11b8d8812cafbafecc6f9239&vid=dea267e876203eb2c2674787701ec602da1b3cf6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Assay Min. 99.0%
Acidity Max. 0.0007 meq/g
Boiling point 67 - 69 °C
Colouration Max. 10 APHA
Density (20/20) 0.723 - 0.728
Evaporation residue Max. 50 ppm
Peroxides (as H2O2) Max. 10 ppm
Water Max. 0.05%
Al (Aluminium) Max. 0.5 ppm
B (Boron) Max. 0.2 ppm
Ba (Barium) Max. 0.2 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.2 ppm
Co (Cobalt) Max. 0.2 ppm
Cr (Chromium) Max. 0.2 ppm
Cu (Copper) Max. 0.2 ppm
Fe (Iron) Max. 0.5 ppm
Mg (Magnesium) Max. 0.2 ppm
Mn (Manganese) Max. 0.2 ppm
Ni (Nickel) Max. 0.2 ppm
Pb (Lead) Max. 0.2 ppm
Sn (Tin) Max. 0.1 ppm
Zn (Zinc) Max. 0.2 ppm
Conforms to ACS Passes Test
Conforms to Reag. Ph. Eur. Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR