Etylenglykol ≥98%, TECHNICAL

Leverantör: VWR Chemicals
Varning

Synonyms: 1,2-Etandiol

24407.292EA 450 SEK
24407.292 24407.326 24407.361 24407.463
Etylenglykol ≥98%, TECHNICAL
Etylenglykol
Formel: HOCH₂CH₂OH
MW: 62.07 g/mol
Kokpunkt: 196…198 °C (1013 hPa)
Smältpunkt: –13 °C
Densitet: 1.115 g/cm³ (20 °C)
Flampunkt: 116
Storage Temperature: Omgivande
MDL Number: MFCD00002885
CAS nummer: 107-21-1
EINECS: 203-473-3
REACH: 01-2119456816-28
Merck Index: 13,03832
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d1c97392851ba184f445f6ee912f84631f98d9&vid=2002300f174929a030d66d9f1394eb6bee89b7dc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Assay (on anhydrous substance) Min. 98 %
Appearance of solution (50 % V/V; water) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR