Järn(II)klorid tetrahydrat 99, purified

Leverantör: VWR Chemicals
Fara

Synonyms: Ferrous chloride tetrahydrate, Iron dichloride tetrahydrate

24128.297EA 1475 SEK
24128.297
Järn(II)klorid tetrahydrat 99, purified
Järn(II)klorid tetrahydrat
Formel: FeCl₂·4H₂O
MW: 198.81 g/mol
Smältpunkt: 677
Densitet: 1.926 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149709
CAS nummer: 13478-10-9
EINECS: 231-843-4
UN: 3260
ADR: 8 III

Order Now

Specifikation testresultat

Assay Min. 99 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR