N,N-Dimetylformamid 99,5, GPR RECTAPUR®

Leverantör: VWR Chemicals
Fara

Synonyms: DMF

23470.293EA 910 SEK
23470.293 23470.327 23470.442 23470.930
N,N-Dimetylformamid 99,5, GPR RECTAPUR®
N,N-Dimetylformamid
Formel: HCON(CH₃)₂
MW: 73.09 g/mol
Kokpunkt: 153
Smältpunkt: –60,5 °C
Densitet: 0.949 g/cm³ (20 °C)
Flampunkt: 57.5 °C (DIN 51755) (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00003284
CAS nummer: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3 III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Assay Min. 99.5 %
Appearance Clear colourless liquid
IR Spectrum Passes test
Acidity Max. 0.00025 meq/g
Alkalinity Max. 0.003 meq/g
Boiling point 152 - 155 °C
Density (20/4) 0.940 - 0.950
Refractive index (20°C) 1.429 - 1.431
Evaporation residue Max. 100 ppm
Water Max. 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR