Diklormetan 99 Stabilised, GPR RECTAPUR®

Leverantör: VWR Chemicals
Varning

Synonyms: Metylenklorid

Stabilization: 20 ppm 2-Methyl-2-buten

23367.296EA 435 SEK
23367.296 23367.321 23367.365 23367.467 23367.550
Diklormetan 99 Stabilised, GPR RECTAPUR®
Diklormetan
Formel: CH₂Cl₂
MW: 84.93 g/mol
Kokpunkt: 39.8 °C (1013 hPa)
Smältpunkt: –95 °C
Densitet: 1.322 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00000881
CAS nummer: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1 III
REACH: 01-2119480404-41
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Assay Min. 99 %
IR Spectrum Passes test
Boiling point 39.0 - 40.5 °C
Acidity Max. 0.0004 meq/g
Density (20/4) 1.322 - 1.327
Evaporation residue Max. 10 ppm
Free chlorine Max. 0.3 ppm
Heavy metals (as Pb) Max. 2 ppm
Water Max. 0.02 %
Cl (Chloride) Max. 3 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR