Slang, Versilon® 2001
Leverantör: Saint Gobain Life Sciences
AE300003 AE300007 AE300012 AE300017 AE300022 AE300027 AE300038 AE300046 AE300053 AE300064
228-1268EA 2600 SEK
228-1268 228-1269 228-1270 228-1271 228-1272 228-1273 228-1274 228-1275 228-1276 228-1277
Slang, Versilon® 2001
Slangar Överföringsslang
Versilon®, clear, for peristaltic pumps.
  • Plasticiser and oil free, does not contaminate fluids
  • Superior flex life in peristaltic pumps
  • Chemically resistant to a wide range of fluids
  • Clear for easy visual flow monitoring
Versilon® 2001 is uniquely engineered to provide flexibility, chemical resistance and extended pump life in a clear, plasticizer/oil-free tubing product. It is resistant to a wide range of fluids that typically destroy PVC products. Its chemical resistance allows it to be used in applications with aggressive chemicals such as polarMore Product Information