Aktivt kol, powder, GPR RECTAPUR®

Leverantör: VWR Chemicals

Synonyms: Carbon activated, Activated carbon, Activated charcoal

22637.293EA 2825 SEK
22637.293 22637.415
Aktivt kol, powder, GPR RECTAPUR®
Aktivt kol
Formel: C
MW: 12.01 g/mol
Smältpunkt: 3550
Densitet: 2 g/cm³ (20 °C)
MDL Number: MFCD00133992
CAS nummer: 64365-11-3
EINECS: 231-153-3
UN: 1362
ADR: 4.2 III
REACH: 01-2119488894-16
Merck Index: 13,01818

Order Now

Specifikation testresultat

Mean particle size Max. 80 µm
Ignition residue Max. 6 %
Water Max. 10 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR