Behållare, bred öppning, kvadratisk, gängad
Leverantör: Kautex
310-84412 310-84419 310-84422 310-84423 310-84593 310-84596 310-84599 310-84601
216-1187EA 960 SEK
216-1187 216-1188 216-1189 216-1200 216-1208 216-1219 216-1220 216-1221
Behållare, bred öppning, kvadratisk, gängad
Provbehållare/Provburk Behållare, allmän användning
PVC. Square. Transparent or brown transparent colours.
  • Wide neck opening
  • Easy to fill and to clean
  • Leakproof
  • Suitable for food
  • With tamper-evident closure, PP, black colour
  • Halal certified
Brown transparent containers are suitable for light-sensitive media. Due to the wide neck opening the containers are easy to fill and to clean. The contents are safe against unauthorized removal. Opening the closure leaves distinct traces.