n-Kapronsyra, Supelco®

Leverantör: Merck
Fara

Synonyms: Hexanoic acid

M-Clarity™ quality level = MQ100

21529-5ML
21529-5MLEA 1380 SEK
21529-5ML
n-Kapronsyra, Supelco®
n-Kapronsyra
Formel: C₆H₁₂O₂
MW: 116.16 g/mol
Kokpunkt: 202
Smältpunkt: –4.8 °C
Densitet: 0.9276 g/cm³ (20 °C)
Flampunkt: 102
Storage Temperature: Omgivande
MDL Number: MFCD00004421
CAS nummer: 142-62-1
EINECS: 205-550-7
UN: 2829
ADR: 8 III
Merck Index: 13,01765
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ec603c3ed7474774d52ad77c6b5ee06d2d921a1&vid=9ecc1dc8f9a2b1255f79e258ca9932e0cb8d2b15&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR