Centrifuge tubes, with screw cap
Leverantör: GLASWARENFABRIK KARL HECHT
40970010
212-0733EA 2025 SEK
212-0733
Centrifuge tubes, with screw cap
Rör Centrifugrör
AR®-Glass
  • Ungraduated
  • Good chemical resistance
Conical bottom, with thread GL 14 and screw cap of polypropylene.