Etanol 96% (v/v), AnalaR NORMAPUR® analytiskt reagens

Leverantör: VWR Chemicals
Fara

Synonyms: Ethyl alcohol

20823.293EA 264 SEK
20823.293 20823.327 20823.362 20823.555 20823.460 20823.290 20823.930
Etanol 96% (v/v), AnalaR NORMAPUR® analytiskt reagens
Etanol

Observera: WARNING : this alcohol is regulated. Call us.
For duty paid orders in the UK please add DP to the product code.

Formel: C₂H₅OH
MW: 46.07 g/mol
Kokpunkt: 78.3 °C (1013 hPa)
Smältpunkt: –117 °C
Densitet: 0.7895 g/cm³ (20 °C)
Flampunkt: 12 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00003568
CAS nummer: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3 II
REACH: 01-2119457610-43
Merck Index: 12,03806

Order Now

Specifikation testresultat

Assay (on anhydrous substance) Min. 99.8 %
Assay (V/V) 95.1 - 96.9 %
IR Spectrum Passes test
UV Spectrum Passes test
Acidity Max. 0.0002 meq/g
Alkalinity Max. 0.0001 meq/g
Boiling point 78.0 - 78.5 °C
Colouration Max. 10 APHA
Substances coloured by H2SO4 Max. 20 APHA
Aldehydes (as CH3CHO) Max. 5 ppm
Aldehydes + ketones (as CH3CHO) Max. 10 ppm
Benzene Max. 5 ppm
Evaporation residue Max. 5 ppm
Formaldehyde Max. 1 ppm
Furaldehyde Max. 1 ppm
Ketones (as CH3COCH3) Max. 10 ppm
Methanol Max. 0.02 %
Propan-2-ol Max. 100 ppm
Substances reducing KMnO4 (as O) Max. 4 ppm
Total heavy alcohols Max. 100 ppm
Water 5.2 - 8.2 %
Al (Aluminium) Max. 0.1 ppm
Ba (Barium) Max. 0.02 ppm
Ca (Calcium) Max. 0.2 ppm
Cd (Cadmium) Max. 0.02 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.01 ppm
Cu (Copper) Max. 0.1 ppm
Fe (Iron) Max. 0.05 ppm
K (Potassium) Max. 0.1 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0.01 ppm
Na (Sodium) Max. 1 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.05 ppm
Sn (Tin) Max. 0.05 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR