Methyl malonyl chloride ≥97%

Leverantör: Thermo Fisher Scientific
Fara

Synonyms: Methyl 3-chloro-3-oxopropanoate

20823.06 20823.14
20823.06EA 1125 SEK
20823.06 20823.14
Methyl malonyl chloride ≥97%
Methyl malonyl chloride

Purity: 97% CAS No.: 37517-81-0 Molecular Formula: C4H5ClO3 Formula Weight: 136.53 Flash Point: 80°(176°F) Physical Form: Liquid Density: 1.27 Refractive Index: 1.4325 Sensitivity: Moisture Sensitive Appearance: Colourless Boiling Point: 58°/12mm UN No.: 3265 MDL No.: MFCD00013657

Formel: C₄H₅ClO₃
MW: 136.53 g/mol
Kokpunkt: 58 °C (12 mmHg)
Densitet: 1.27
Flampunkt: 80 °C (176 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00013657
CAS nummer: 37517-81-0
EINECS: 253-540-6
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR