Svavelsyra 50%, TECHNICAL

Leverantör: VWR Chemicals
Fara

20696.293EA 375 SEK
20696.293 20696.460
Svavelsyra 50%, TECHNICAL
Svavelsyra
Formel: H₂SO₄
MW: 98.08 g/mol
Kokpunkt: >100 °C (1013 hPa)
Smältpunkt: 10,38
Densitet: 1.1…1.4 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00064589
CAS nummer: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II

Order Now

Specifikation testresultat

Assay Min. 50 %

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR