Bärnstenssyra ≥99.5%, AnalaR NORMAPUR® analytiskt reagens

Leverantör: VWR Chemicals
Varning

Synonyms: 1,4-Butanedioic acid

20668.230EA 660 SEK
20668.230 20668.410
Bärnstenssyra ≥99.5%, AnalaR NORMAPUR® analytiskt reagens
Bärnstenssyra
Formel: HOOCCH₂CH₂COOH
MW: 118.09 g/mol
Kokpunkt: 235
Smältpunkt: 185-191 °C
Densitet: 1.56 g/cm³ (20 °C)
Flampunkt: 206
Storage Temperature: Omgivande
MDL Number: MFCD00002789
CAS nummer: 110-15-6
EINECS: 203-740-4
UN: 3261
ADR: 8 III
Merck Index: 13,08953
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfd381e6e0e4cbde676a92c9e8d6aacfac635ade&vid=0b14e1b7f82b03468fc82d224a0ee894952eb200&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Assay Min. 99.5 %
Melting point 185-191 °C
Heavy metals (as Pb) Max. 10 ppm
Ignition residue (SO4) Max. 0.02 %
Insolubility in water Max. 50 ppm
Water Max. 0.5 %
Cl (Chloride) Max. 5 ppm
NH4 (Ammonium) Max. 10 ppm
SO4 (Sulphate) Max. 50 ppm
Fe (Iron) Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR